349763
3-(3-Methoxyphenyl)propionic acid
99%
Synonym(s):
3-Methoxyhydrocinnamic acid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Assay
99%
form
powder
mp
43-45 °C (lit.)
SMILES string
COc1cccc(CCC(O)=O)c1
InChI
1S/C10H12O3/c1-13-9-4-2-3-8(7-9)5-6-10(11)12/h2-4,7H,5-6H2,1H3,(H,11,12)
InChI key
BJJQJLOZWBZEGA-UHFFFAOYSA-N
General description
3-(3-Methoxyphenyl)propionic acid can be prepared by the catalytic (palladium on charcoal) reduction of its corresponding unsaturated acid with H2.
Application
3-(3-Methoxyphenyl)propionic acid may be used in chemical synthesis.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Applied and environmental microbiology, 44(3), 597-603 (1982-09-01)
Isolated rumen bacteria were examined for growth and, where appropriate, for their ability to degrade cellulose in the presence of the hydroxycinnamic acids trans-p-coumaric acid and trans-ferulic acid and the hydroxybenzoic acids vanillic acid and 4-hydroxybenzoic acid. Ferulic and p-coumaric
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service