Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

246123

Sigma-Aldrich

5-Isoquinolinesulfonic acid

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H7NO3S
CAS Number:
Molecular Weight:
209.22
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

powder

impurities

2-5% H2SO4

mp

>300 °C (lit.)

functional group

sulfonic acid

SMILES string

OS(=O)(=O)c1cccc2cnccc12

InChI

1S/C9H7NO3S/c11-14(12,13)9-3-1-2-7-6-10-5-4-8(7)9/h1-6H,(H,11,12,13)

InChI key

YFMJTLUPSMCTOQ-UHFFFAOYSA-N

Application

5-Isoquinolinesulfonic acid was used in the synthesis of conjugates of oligoarginine peptides[1].

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Erki Enkvist et al.
Journal of medicinal chemistry, 49(24), 7150-7159 (2006-11-28)
Conjugates of oligoarginine peptides with adenine, adenosine, adenosine-5'-carboxylic acid, and 5-isoquinolinesulfonic acid were synthesized and characterized as bisubstrate-analog inhibitors of cAMP-dependent protein kinase. Adenosine and adenine derivatives were connected to the N- or C-terminus of peptides containing four to six

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service