MilliporeSigma
  • Conjugation of adenosine and hexa-(D-arginine) leads to a nanomolar bisubstrate-analog inhibitor of basophilic protein kinases.
Conjugation of adenosine and hexa-(D-arginine) leads to a nanomolar bisubstrate-analog inhibitor of basophilic protein kinases.
Journal of medicinal chemistry (2006-11-28)
Erki Enkvist, Darja Lavogina, Gerda Raidaru, Angela Vaasa, Indrek Viil, Marje Lust, Kaido Viht, Asko Uri
ABSTRACT
Conjugates of oligoarginine peptides with adenine, adenosine, adenosine-5'-carboxylic acid, and 5-isoquinolinesulfonic acid were synthesized and characterized as bisubstrate-analog inhibitors of cAMP-dependent protein kinase. Adenosine and adenine derivatives were connected to the N- or C-terminus of peptides containing four to six L- or D-arginine residues via a linker with a length that had been optimized in structure-activity studies. The orientation of the peptide chain strongly affected the activity of compounds incorporating D-arginines. The biligand inhibitor containing Hidaka's H9 isoquinolinesulfonamide connected to the L-peptide had 65 times higher potency than the corresponding adenosine-containing conjugate, while both types of the conjugate comprising D-peptides had similar low nanomolar activity. Two of the most active adenosine- and H9-peptide conjugates were tested in the panel of 52 different kinases. At 1 microM concentration, both compounds showed strong (more than 95%) inhibition of several basophilic AGC kinases, including pharmaceutically important kinases ROCK II and PKB/Akt.