Skip to Content
MilliporeSigma
All Photos(1)

Documents

216755

Sigma-Aldrich

5-Nitroindazole

≥99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H5N3O2
CAS Number:
Molecular Weight:
163.13
Beilstein:
7936
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

≥99%

form

powder

mp

204-208 °C (lit.)

SMILES string

[O-][N+](=O)c1ccc2[nH]ncc2c1

InChI

1S/C7H5N3O2/c11-10(12)6-1-2-7-5(3-6)4-8-9-7/h1-4H,(H,8,9)

InChI key

WSGURAYTCUVDQL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

5-Nitroindazole inhibits citrulline formation by Ca2+-calmodulin (CaM)-dependent nitric oxide synthase from bovine brain. Nitration of 5-nitroindazole with nitric acid and acetic anhydride yields 3,5-dintroindazole and 2,5-dinitroindazole.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nitration of indazoles in the 3 position.
Habraken CL and Cohen-Fernandes P.
The Journal of Organic Chemistry, 36(21), 3084-3086 (1971)
D J Wolff et al.
Archives of biochemistry and biophysics, 311(2), 300-306 (1994-06-01)
Citrulline formation by Ca(2+)-calmodulin (CaM)-dependent nitric oxide synthase from bovine brain is inhibited reversibly by indazole, 5-nitro-, 6-nitro-, and 7-nitroindazole with IC50 values of 2.3 mM, 1.15 mM, 40 microM, and 2.5 microM, respectively. Inhibition of citrulline formation by 7-nitroindazole

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service