Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

163937

Sigma-Aldrich

N,N,N′,N′-Tetramethylphosphorodiamidic chloride

technical grade, 90%

Synonym(s):

Bis(dimethylamido)phosphoric chloride

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$85.80
50 G
$249.00

$85.80


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 G
$85.80
50 G
$249.00

About This Item

Linear Formula:
[(CH3)2N]2P(O)Cl
CAS Number:
Molecular Weight:
170.58
Beilstein:
471430
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$85.80


Please contact Customer Service for Availability

Request a Bulk Order

grade

technical grade

Quality Level

Assay

90%

form

liquid

refractive index

n20/D 1.466 (lit.)

density

1.152 g/mL at 25 °C (lit.)

functional group

phosphoramide

SMILES string

CN(C)P(Cl)(=O)N(C)C

InChI

1S/C4H12ClN2OP/c1-6(2)9(5,8)7(3)4/h1-4H3

InChI key

WYLQARGYFXBZMD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

N,N,N′,N′-Tetramethylphosphorodiamidic chloride was used in the synthesis of (-)-allohedycaryol[1] and C2-symmetric binaphthyl phosphortriamide[2]. It was also used in total synthesis of neohedycaryol[3].

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A I Meyers et al.
Chirality, 9(5-6), 431-434 (1997-01-01)
By use of an asymmetric Ullmann coupling involving chiral naphthalene oxazolines 1, the title compounds were prepared in good yields and with high diastereoselectivity. Hydrolysis of the binaphthyl oxazolines 2 led to the di-aldehydes 5, which were transformed into the
Vladimir N. Zhabinskii et al.
The Journal of organic chemistry, 61(12), 4022-4027 (1996-06-14)
The enantiomer of (+)-allohedycaryol, a germacrane alcohol isolated from giant fennel (Ferula communis L.), has been synthesized, thereby elucidating the relative and absolute stereochemistry of the natural product. The synthesis of (-)-allohedycaryol started from (+)-alpha-cyperone (5) which was available in
Adriaan J. Minnaard et al.
The Journal of organic chemistry, 62(8), 2344-2349 (1997-04-18)
The total synthesis of neohedycaryol (4), the C(9)-C(10) double bond regioisomer of the germacrane sesquiterpene hedycaryol, was accomplished in 10 steps from the known dione 6. A Marshall fragmentation of the intermediate mesylate 14 was used to prepare the trans,trans-cyclodeca-1,6-diene

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service