MilliporeSigma
  • Relative and Absolute Configuration of Allohedycaryol. Enantiospecific Total Synthesis of Its Enantiomer.
Relative and Absolute Configuration of Allohedycaryol. Enantiospecific Total Synthesis of Its Enantiomer.
The Journal of organic chemistry (1996-06-14)
Vladimir N. Zhabinskii, Adriaan J. Minnaard, Joannes B. P. A. Wijnberg, Aede de Groot
ABSTRACT
The enantiomer of (+)-allohedycaryol, a germacrane alcohol isolated from giant fennel (Ferula communis L.), has been synthesized, thereby elucidating the relative and absolute stereochemistry of the natural product. The synthesis of (-)-allohedycaryol started from (+)-alpha-cyperone (5) which was available in relatively large quantities via alkylation of imine 7 derived from (+)-dihydrocarvone and (R)-(+)-1-phenylethylamine. In a number of steps 5 was converted into the mesylate 4with a regio- and stereoselective epoxidation as the key step. A Marshall fragmentation of 4 was used to prepare the trans,trans-cyclodeca-1,6-diene ring present in allohedycaryol. The conformation of synthetic (-)-allohedycaryol was elucidated via photochemical conversion into a bourbonane system. The synthesis of (-)-allohedycaryol also showed that natural (+)-allohedycaryol has the opposite absolute stereochemistry to that normally found in higher plants.
Sigma-Aldrich
N,N,N′,N′-Tetramethylphosphorodiamidic chloride, technical grade, 90%
Pricing and availability is not currently available.