MilliporeSigma
  • Synthesis and chiroptical properties of a novel C2-symmetric binaphthyl phosphortriamide ("chiral HMPA").
Synthesis and chiroptical properties of a novel C2-symmetric binaphthyl phosphortriamide ("chiral HMPA").
Chirality (1997-01-01)
A I Meyers, T Nguyen, D Stoianova, N Sreerama, R W Woody, A Koslowski, J Fleischhauer
ABSTRACT
By use of an asymmetric Ullmann coupling involving chiral naphthalene oxazolines 1, the title compounds were prepared in good yields and with high diastereoselectivity. Hydrolysis of the binaphthyl oxazolines 2 led to the di-aldehydes 5, which were transformed into the azepine derivative, 6. The latter was treated with the appropriate phosphoryl halide to access the chiral HMPA systems 7 and 9. The CD spectra of the chiral azepine 6 and the chiral phosphoramides 7 and 9 were measured and showed a strong positive CD couplet near 225 nm, consistent with the P axial chirality (S configuration). Semi-empirical CNDO/S molecular orbital calculations of the CD spectrum of 6 satisfactorily reproduced the major features of the observed spectrum.
Sigma-Aldrich
N,N,N′,N′-Tetramethylphosphorodiamidic chloride, technical grade, 90%