Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

152986

Sigma-Aldrich

1-Benzyl-4-hydroxypiperidine

96%

Synonym(s):

1-Benzyl-4-piperidinol

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$136.00

$136.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$136.00

About This Item

Empirical Formula (Hill Notation):
C12H17NO
CAS Number:
Molecular Weight:
191.27
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$136.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

96%

form

solid

bp

127-128 °C/2 mmHg (lit.)

mp

61-63 °C (lit.)

functional group

hydroxyl
phenyl

SMILES string

OC1CCN(CC1)Cc2ccccc2

InChI

1S/C12H17NO/c14-12-6-8-13(9-7-12)10-11-4-2-1-3-5-11/h1-5,12,14H,6-10H2

InChI key

BPPZXJZYCOETDA-UHFFFAOYSA-N

Application

1-Benzyl-4-hydroxypiperidine was used as an alternative molecule to study the ligand concentration attached to the epoxy-activated Sepharose 6B[1].
Reactant for synthesis of:
Muscarinic acetylcholine receptor antagonist and beta 2 adrenoceptor agonist
Fatty acid amide hydrolase inhibitors
PI3 kinase-alpha inhibitors
Flavonoid derivatives used as dual binding acetylcholinesterase inhibitors
Urotensin-II receptor antagonists
Rho kinase inhibitors

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

X Santarelli et al.
Journal of chromatography. B, Biomedical sciences and applications, 739(1), 63-72 (2000-04-01)
New pseudo-affinity chromatographic supports for penicillin acylase were prepared and evaluated with three different samples: pure penicillin acylase, industrial clarified feedstock and crude extract. The different gels were studied for their purification fold (three to six) and their recovery power

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service