Skip to Content
MilliporeSigma
All Photos(2)

Documents

H42206

Sigma-Aldrich

N-Methyl-4-piperidinol

98%

Synonym(s):

1-Methyl-4-piperidinol, 4-Hydroxy-1-methylpiperidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H13NO
CAS Number:
Molecular Weight:
115.17
Beilstein:
103024
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.4775 (lit.)

bp

200 °C (lit.)

mp

29-31 °C (lit.)

storage temp.

2-8°C

SMILES string

CN1CCC(O)CC1

InChI

1S/C6H13NO/c1-7-4-2-6(8)3-5-7/h6,8H,2-5H2,1H3

InChI key

BAUWRHPMUVYFOD-UHFFFAOYSA-N

Application

Reactant for:
  • Optimization of Novobiocin scaffold to product antitumor agents
  • Substitution about the rigidifying ring for histamine H4 receptor antagonist synthesis

Reactant for synthesis of:
  • CaMKII inhibitors
  • VEGFR and FGFR kinase inhibitors
  • Phosphoinositide-3-kinase inhibitors
  • Protein lysine methyltransferase G9a inhibitors

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

233.6 °F - closed cup

Flash Point(C)

112 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service