Merck
Molecular activation of PPARgamma by angiotensin II type 1-receptor antagonists.
Vascular pharmacology (2006-06-13)
David V Erbe, Katherine Gartrell, Yan-Ling Zhang, Vipin Suri, Steven J Kirincich, Sarah Will, Mylene Perreault, Suyue Wang, James F Tobin
RESUMO
Elevated blood pressure and insulin resistance are strongly associated in patients. We explored the potential for the anti-hypertensive angiotensin II type 1-receptor (ATR(1)) antagonists to improve insulin sensitivity through modulation of the nuclear receptor PPARgamma, in vitro and in vivo compared to the potent insulin sensitizer, rosiglitazone. PPARgamma modulation by ATR(1) antagonists was measured first by direct recruitment of PGC-1, followed by trans-activation reporter assays in cells, and promotion of adipogenesis in fibroblast and pre-adipocyte cell lines. Improvement of insulin sensitivity was measured as changes in levels of glucose, insulin, and adiponectin in ob/ob mice. Telmisartan, candesartan, irbesartan, and losartan (but not valsartan or olmesartan) each served as bona fide PPARgamma ligands in vitro, with EC(50) values between 3 and 5 micro mol/l. However, only telmisartan, and to a lesser extent candesartan, resulted in significant PPARgamma agonism in cells. In vivo, although rosiglitazone significantly lowered both glucose (33%, p<0.01) and insulin (61%, p<0.01) levels and increased expression of adiponectin (74%, p<0.001), sartan treatment had no effect. Many members of the sartan family of ATR(1) antagonists are PPARgamma ligands in cell-free assays but their modulation of PPARgamma in cells is relatively weak. Furthermore, none appear to improve insulin sensitivity in a rodent model under conditions where other insulin sensitizers, including rosiglitazone, do. These results question whether reported effects of sartans on insulin sensitivity may be through other means, and should guide further efforts to develop dual agents to treat hypertension and insulin resistance.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Irbesartan, ≥98% (HPLC), powder
Millipore
Mouse Adiponectin ELISA, This Mouse Adiponectin ELISA is used to measure & quantify Adiponectin levels in Metabolism & Endocrine research.