Merck
  • Scalable High-Affinity Stabilization of Magnetic Iron Oxide Nanostructures by a Biocompatible Antifouling Homopolymer.
Scalable High-Affinity Stabilization of Magnetic Iron Oxide Nanostructures by a Biocompatible Antifouling Homopolymer.
ACS applied materials & interfaces (2017-10-13)
Giovanni Luongo, Paola Campagnolo, Jose E Perez, Jürgen Kosel, Theoni K Georgiou, Anna Regoutz, David J Payne, Molly M Stevens, Mary P Ryan, Alexandra E Porter, Iain E Dunlop
RESUMO
Iron oxide nanostructures have been widely developed for biomedical applications because of their magnetic properties and biocompatibility. In clinical applications, stabilization of these nanostructures against aggregation and nonspecific interactions is typically achieved using weakly anchored polysaccharides, with better-defined and more strongly anchored synthetic polymers not commercially adopted because of their complexity of synthesis and use. Here, we show for the first time stabilization and biocompatibilization of iron oxide nanoparticles by a synthetic homopolymer with strong surface anchoring and a history of clinical use in other applications, poly(2-methacryloyloxyethyl phosphorylcholine) [poly(MPC)]. For the commercially important case of spherical particles, binding of poly(MPC) to iron oxide surfaces and highly effective individualization of magnetite nanoparticles (20 nm) are demonstrated. Next-generation high-aspect-ratio nanowires (both magnetite/maghemite and core-shell iron/iron oxide) are, furthermore, stabilized by poly(MPC) coating, with the nanowire cytotoxicity at large concentrations significantly reduced. The synthesis approach exploited to incorporate functionality into the poly(MPC) chain is demonstrated by random copolymerization with an alkyne-containing monomer for click chemistry. Taking these results together, poly(MPC) homopolymers and random copolymers offer a significant improvement over current iron oxide nanoformulations, combining straightforward synthesis, strong surface anchoring, and well-defined molecular weight.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
5-Carboxytetramethylrhodamine