- Reciprocal expression of INSM1 and YAP1 defines subgroups in small cell lung cancer.
Reciprocal expression of INSM1 and YAP1 defines subgroups in small cell lung cancer.
Oncotarget (2017-11-02)
Karen McColl, Gary Wildey, Nneha Sakre, Mary Beth Lipka, Mohadese Behtaj, Adam Kresak, Yanwen Chen, Michael Yang, Vamsidhar Velcheti, Pingfu Fu, Afshin Dowlati
PMID29088741
RESUMO
The majority of small cell lung cancer (SCLC) patients demonstrate initial chemo-sensitivity, whereas a distinct subgroup of SCLC patients, termed chemo-refractory, do not respond to treatment. There is little understanding of how to distinguish these patients prior to disease treatment. Here we used gene expression profiling to stratify SCLC into subgroups and characterized a molecular phenotype that may identify, in part, chemo-refractive SCLC patients. Two subgroups of SCLC were identified in both cell lines and tumors by the reciprocal expression of two genes;
MATERIAIS
Número do produto
Marca
Descrição do produto