Merck
  • Mig-6 is down-regulated in HCC and inhibits the proliferation of HCC cells via the P-ERK/Cyclin D1 pathway.
Mig-6 is down-regulated in HCC and inhibits the proliferation of HCC cells via the P-ERK/Cyclin D1 pathway.
Experimental and molecular pathology (2017-05-17)
Zixuan Li, Lianyue Qu, Wenting Luo, Yulong Tian, Huan Zhai, Ke Xu, Hongshan Zhong
RESUMO
The ablation of Mig-6 has been shown to induce tumor formation in various tissues. However, the relationships between Mig-6 expression, clinical pathological factors, and prognosis have not been clarified in hepatocellular carcinoma (HCC), and the mechanism by which Mig-6 regulates the proliferation of HCC cells has not been reported. In this study, we investigated the clinical significance of the loss of Mig-6 expression in HCC and the mechanism underlying the inhibition of cell proliferation by Mig-6. The down-regulation of Mig-6 correlated significantly with large tumors, a more advanced BCLC stage, and a more advanced TNM stage, and low Mig-6 expression predicted significantly reduced survival. Low Mig-6 expression and high Cyclin D1 expression were independent predictors for survival. The overexpression of Mig-6 led to significant G
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
MISSION® esiRNA, targeting human ERRFI1