- Novel screening system for high-affinity ligand of heredity vitamin D-resistant rickets-associated vitamin D receptor mutant R274L using bioluminescent sensor.
Novel screening system for high-affinity ligand of heredity vitamin D-resistant rickets-associated vitamin D receptor mutant R274L using bioluminescent sensor.
The Journal of steroid biochemistry and molecular biology (2016-11-20)
Hiroki Mano, Miyu Nishikawa, Kaori Yasuda, Shinichi Ikushiro, Nozomi Saito, Daisuke Sawada, Shinobu Honzawa, Masashi Takano, Atsushi Kittaka, Toshiyuki Sakaki
PMID27864003
RESUMO
Hereditary vitamin D-resistant rickets (HVDRR) is caused by mutations in the vitamin D receptor (VDR) gene. Arg274 located in the ligand binding domain (LBD) of VDR is responsible for anchoring 1α,25-dihydroxyvitamin D3 (1α,25(OH)
MATERIAIS
Número do produto
Marca
Descrição do produto
Ethanol solution, NIST® SRM® 2899a, nominal 25% by mass