Merck
Glycosides from edible sea cucumbers stimulate macrophages via purinergic receptors.
Scientific reports (2016-12-23)
Dmitry Aminin, Evgeny Pislyagin, Maxim Astashev, Andrey Es'kov, Valery Kozhemyako, Sergei Avilov, Elena Zelepuga, Ekaterina Yurchenko, Leonid Kaluzhskiy, Emma Kozlovskaya, Alexis Ivanov, Valentin Stonik
RESUMO
Since ancient times, edible sea cucumbers have been considered a jewel of the seabed and used in Asian folk medicine for stimulation of resistance against different diseases. However, the power of this sea food has not been established on a molecular level. A particular group of triterpene glycosides was found to be characteristic metabolites of the animals, responsible for this biological action. Using one of them, cucumarioside A2-2 (CA2-2) from the edible Cucumaria japonica species as an example as well as inhibitory analysis, patch-clamp on single macrophages, small interfering RNA technique, immunoblotting, SPR analysis, computer modeling and other methods, we demonstrate low doses of CA2-2 specifically to interact with P2X receptors (predominantly P2X4) on membranes of mature macrophages, enhancing the reversible ATP-dependent Ca2+ intake and recovering Ca2+ transport at inactivation of these receptors. As result, interaction of glycosides of this type with P2X receptors leads to activation of cellular immunity.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
MISSION® esiRNA, targeting human P2RX4