Merck
First triazole-bridged unsymmetrical porphyrin dyad via click chemistry.
The Journal of organic chemistry (2007-12-12)
Sokkalingam Punidha, Jasmine Sinha, Anil Kumar, Mangalampalli Ravikanth
RESUMO
Click chemistry has been successfully applied in the synthesis of the first example of a triazole-bridged porphyrin dyad containing N(2)S(2) porphyrin and N(4) or ZnN(4) porphyrin subunits, and fluorescence study indicated a possibility of singlet-singlet energy transfer from the N(4) or ZnN(4) porphyrin subunit to the N(20S(2) porphyrin subunit.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
3,4-Dimethoxythiophene, 97%