Merck
Beta-catenin regulation during the cell cycle: implications in G2/M and apoptosis.
Molecular biology of the cell (2003-07-15)
David Olmeda, Susanna Castel, Senén Vilaró, Amparo Cano
RESUMO
Beta-catenin is a multifunctional protein involved in cell-cell adhesion and Wnt signal transduction. Beta-catenin signaling has been proposed to act as inducer of cell proliferation in different tumors. However, in some developmental contexts and cell systems beta-catenin also acts as a positive modulator of apoptosis. To get additional insights into the role of beta-catenin in the regulation of the cell cycle and apoptosis, we have analyzed the levels and subcellular localization of endogenous beta-catenin and its relation with adenomatous polyposis coli (APC) during the cell cycle in S-phase-synchronized epithelial cells. Beta-catenin levels increase in S phase, reaching maximum accumulation at late G2/M and then abruptly decreasing as the cells enter into a new G1 phase. In parallel, an increased cytoplasmic and nuclear localization of beta-catenin and APC is observed during S and G2 phases. In addition, strong colocalization of APC with centrosomes, but not beta-catenin, is detected in M phase. Interestingly, overexpression of a stable form of beta-catenin, or inhibition of endogenous beta-catenin degradation, in epidermal keratinocyte cells induces a G2 cell cycle arrest and leads to apoptosis. These results support a role for beta-catenin in the control of cell cycle and apoptosis at G2/M in normal and transformed epidermal keratinocytes.
MATERIAIS
Número do produto
Marca
Descrição do produto
Roche
RNase, from bovine pancreas