Merck
Fully automated continuous flow synthesis of 4,5-disubstituted oxazoles.
Organic letters (2006-11-03)
Marcus Baumann, Ian R Baxendale, Steven V Ley, Christoper D Smith, Geoffrey K Tranmer
RESUMO
[Structure: see text] A multipurpose mesofluidic flow reactor capable of producing gram quantities of material has been developed as an automated synthesis platform for the rapid on-demand synthesis of key building blocks and small exploratory libraries. The reactor is configured to provide the maximum flexibility for screening of reaction parameters that incorporate on-chip mixing and columns of solid supported reagents to expedite the chemical syntheses.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Ethyl isocyanoacetate, 95%