Merck
New synthetic substrates of mammalian nucleotide excision repair system.
Nucleic acids research (2013-04-24)
Alexey Evdokimov, Irina Petruseva, Aleksandra Tsidulko, Ludmila Koroleva, Inna Serpokrylova, Vladimir Silnikov, Olga Lavrik
RESUMO
DNA probes for the studies of damaged strand excision during the nucleotide excision repair (NER) have been designed using the novel non-nucleosidic phosphoramidite reagents that contain N-[6-(9-antracenylcarbamoyl)hexanoyl]-3-amino-1,2-propandiol (nAnt) and N-[6-(5(6)-fluoresceinylcarbamoyl)hexanoyl]-3-amino-1,2-propandiol (nFlu) moieties. New lesion-imitating adducts being inserted into DNA show good substrate properties in NER process. Modified extended linear nFlu- and nAntr-DNA are suitable for estimation of specific excision activity catalysed with mammalian whole-cell extracts. The following substrate activity range was revealed for the model 137-bp linear double-stranded DNA: nAnt-DNA ≈ nFlu-DNA > Chol-DNA (Chol-DNA--legitimate NER substrate that contains non-nucleoside fragment bearing cholesterol residue). In vitro assay shows that modified DNA can be a useful tool to study NER activity in whole-cell extracts. The developed approach should be of general use for the incorporation of NER-sensitive distortions into model DNAs. The new synthetic extended linear DNA containing bulky non-nucleoside modifications will be useful for NER mechanism study and for applications.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Trimethylacetic anhydride, 99%
Preço e disponibilidade não estão disponíveis no momento.