Merck
  • Trisubstituted 2-trifluoromethyl pyrrolidines via catalytic asymmetric Michael addition/reductive cyclization.
Trisubstituted 2-trifluoromethyl pyrrolidines via catalytic asymmetric Michael addition/reductive cyclization.
Organic letters (2014-04-22)
Michael T Corbett, Qihai Xu, Jeffrey S Johnson
RESUMO
The stereoselective synthesis of trisubstituted 2-trifluoromethyl pyrrolidines by asymmetric Michael addition/hydrogenative cyclization is described. The direct organocatalytic addition of 1,1,1-trifluoromethylketones to nitroolefins proceeds under mild reaction conditions and low catalyst loadings to provide Michael adducts in high yield with excellent diastereo- and enantioselectivity. Catalytic hydrogenation of the Michael adducts stereoselectively generates 2-trifluoromethylated pyrrolidines bearing three contiguous stereocenters. A stereospecific route to epimeric 2-trifluoromethyl pyrrolidines from a common intermediate is described.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Schreiner′s Thiourea Catalyst, 95%