Merck
PAI-1 mediates the antiangiogenic and profibrinolytic effects of 16K prolactin.
Nature medicine (2014-06-16)
Khalid Bajou, Stephanie Herkenne, Victor L Thijssen, Salvino D'Amico, Ngoc-Quynh-Nhu Nguyen, Ann Bouché, Sébastien Tabruyn, Mohammed Srahna, Jean-Yves Carabin, Olivier Nivelles, Cécile Paques, Ivo Cornelissen, Michelle Lion, Agnès Noel, Ann Gils, Stefan Vinckier, Paul J Declerck, Arjan W Griffioen, Mieke Dewerchin, Joseph A Martial, Peter Carmeliet, Ingrid Struman
RESUMO
The N-terminal fragment of prolactin (16K PRL) inhibits tumor growth by impairing angiogenesis, but the underlying mechanisms are unknown. Here, we found that 16K PRL binds the fibrinolytic inhibitor plasminogen activator inhibitor-1 (PAI-1), which is known to contextually promote tumor angiogenesis and growth. Loss of PAI-1 abrogated the antitumoral and antiangiogenic effects of 16K PRL. PAI-1 bound the ternary complex PAI-1-urokinase-type plasminogen activator (uPA)-uPA receptor (uPAR), thereby exerting antiangiogenic effects. By inhibiting the antifibrinolytic activity of PAI-1, 16K PRL also protected mice against thromboembolism and promoted arterial clot lysis. Thus, by signaling through the PAI-1-uPA-uPAR complex, 16K PRL impairs tumor vascularization and growth and, by inhibiting the antifibrinolytic activity of PAI-1, promotes thrombolysis.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Prolactin human, recombinant, expressed in E. coli, lyophilized powder, BioReagent, suitable for cell culture, >97% (SDS-PAGE)