Merck
  • Vibrational analysis and valence force field for nitrotoluenes, dimethylanilines and some substituted methylbenzenes.
Vibrational analysis and valence force field for nitrotoluenes, dimethylanilines and some substituted methylbenzenes.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2012-08-08)
Jai Kishan Ojha, B Venkatram Reddy, G Ramana Rao
RESUMO
The Fourier transform infrared (FTIR) and Raman spectra of 2-amino-4-nitrotoluene; 2-amino-5-nitrotoluene; 2,4-dimethylaniline; 2,5-dimethylaniline; 2,6-dimethylaniline; 1,2,4-trimethylbenzene; 1,3,5-trimethylbenzene and pentamethylbenzene have been recorded in the range 4000-400 cm(-1) and 4000-30 cm(-1), respectively. A normal coordinate analysis was carried out for both in-plane and out-of-plane vibrations of these molecules using an 81-parameter modified valence force field. The force constants were refined using 369 frequencies of eight molecules in the overlay least-squares technique. The reliability of the force constants was tested by making a zero-order calculation for both in-plane and out-of-plane vibrations for five related molecules. The potential energy distributions and eigen vectors calculated in the process were used to make unambiguous vibrational assignments of all the fundamentals.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
2,5-Dimethylaniline, 99%