Merck
  • Synthesis and evaluation of antitumor activity of novel 2-[Nmethyl-N-(4-methyl-1,3-benzothiazol-2-yl)aminomethyl]-5,8-diacyloxy-1,4-naphthoquinones.
Synthesis and evaluation of antitumor activity of novel 2-[Nmethyl-N-(4-methyl-1,3-benzothiazol-2-yl)aminomethyl]-5,8-diacyloxy-1,4-naphthoquinones.
Archives of pharmacal research (2008-03-28)
Jikang Yoo, Hyun-Suk Choi, Cheol-Hee Choi, Yongseog Chung, Bok Hee Kim, Hoon Cho
RESUMO
A series of nine new compounds bridged by acyl groups at the 5,8-dihydroxyl group of DHNQ were synthesized and their cytotoxic activity against L1210 and P388 cancer cells was examined. Their antitumor action in mice bearing S-180 cells in the peritoneal cavity was also assessed. Increasing the size of the acyl group (compounds 7-9) up to propyl increased the antitumor activity (T/C value), whereas the cytotoxicity of these compounds was comparable against L1210 (lymphocytic leukemia) and P388 (lymphoid neoplasm) cancer cells. Further increasing in the chain length (compounds 11-15) decreased the potency. Thus, acyl group chains of three carbon atoms is optimal for antitumor activity. The most potent compound of this series was 2-[N-methyl-N-(4-methyl-1,3-benzothiazol-2-yl)aminomethyl]-5,8-dipropylcarbonyloxy-1,4-naphthoquinone (compound 9) with a T/C (%) value of 354.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
2-Amino-4-methylbenzothiazole, 97%