Merck
  • Observation and elimination of N-acetylation of oligonucleotides prepared using fast-deprotecting phosphoramidites and ultra-mild deprotection.
Observation and elimination of N-acetylation of oligonucleotides prepared using fast-deprotecting phosphoramidites and ultra-mild deprotection.
Bioorganic & medicinal chemistry letters (2001-05-17)
Q Zhu, M O Delaney, M M Greenberg
RESUMO
Commercially available 'fast-deprotecting' phosphoramidites are useful for synthesizing oligonucleotides containing alkali-sensitive nucleotides. However, N-acetylated oligonucleotides were observed during solid-phase synthesis using 'fast-deprotecting' phosphoramidites in conjunction with K2CO3/MeOH ('ultra-mild') deprotection. Transamidation was localized at deoxyguanosine, which is protected as its isopropylphenoxyacetyl amide. Substitution of trimethylacetic anhydride for acetic anhydride and appropriate modification of the automated synthesis cycles eliminated this problem.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Trimethylacetic anhydride, 99%