Merck
  • Structural properties, cytotoxicity, and anti-inflammatory activity of silver(I) complexes with tris(p-tolyl)phosphine and 5-chloro-2-mercaptobenzothiazole.
Structural properties, cytotoxicity, and anti-inflammatory activity of silver(I) complexes with tris(p-tolyl)phosphine and 5-chloro-2-mercaptobenzothiazole.
Bioinorganic chemistry and applications (2010-04-10)
L Kyros, N Kourkoumelis, M Kubicki, L Male, M B Hursthouse, I I Verginadis, E Gouma, S Karkabounas, K Charalabopoulos, S K Hadjikakou
RESUMO
The synthesis and characterization of the silver(I) chloride complex of formula {[AgCI(CMBZT)(TPTP)(2)] . (MeOH)} (1) (CMBZT = 5-chloro-2-mercaptobenzothiazole, TPTP = tris(p-tolyl)phosphine) is described. Also the structure of the hydrate derivative {[AgCI(TPTP)(3)] . (0.5 . H(2)O)} (2) of the corresponding known anhydrous silver complex (Zartilas et al., 2009), and the polymorph 3 of the known [AgI(TPTP)(3)] complex (Zartilas et al., 2009) were determined and compared with the known ones. In addition, the structure of the known one silver(I) cluster {[AgI(TPTP)](4)} (4) (Meijboom et al., 2009) was re-determined at 120(2) K and possible Ag-Ag interactions were analyzed. The compounds 1-4 were characterized by X-ray crystallography at r.t (1) and 120 K (2-4). All these complexes and {[(Et(3)NH)(+)](2) . [Ag(6)(mu(3)-Hmna)(4)(mu(3)-mna)(2)](2-) . (DMSO)(2) . (H(2)O)} (5) (Hmna = 2-mercaptonicotinic acid) were evaluated for cytotoxic and anti-inflammatory activity. The in vitro testing of cytotoxic activity of 1-5 against leiomyosarcoma cancer cells (LMS), were evaluated with Trypan Blue and Thiazolyl Blue Tetrazolium Bromide or 3-(4.5-dimethylthiazol-2-yl)-2.5-diphenyltetrazolium bromide (MTT) assays. The flow cytometry assay for complex 1 and showed that at 15 muM of 1, 62.38% of LMS cells undergo apoptosis, while 7% of LMS cells undergo cell necrosis. The antitumor activity of 3 is comparable with that of its reported polymorph (Zartilas et al., 2009). The anti-inflammatory, activity of complexes 1-3 and 5 was also studied. The activity towards cell viability was 2 > 3 > 5 > 1 > 4, while the order of the inhibitory activity in cell growth proliferation follows the order, 2 > 3 > 1 > 4 > 5. The anti-inflammatory activity on the other hand is 1 > 2 > 5 > cdots, three dots, centered >3.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
5-Chloro-2-mercaptobenzothiazole, technical grade, ≥90%