- Octatropine-methyl-bromide and sulglycotide salt in the short-term treatment of active duodenal ulcer. A double blind endoscopic study of 40 outpatients.
Octatropine-methyl-bromide and sulglycotide salt in the short-term treatment of active duodenal ulcer. A double blind endoscopic study of 40 outpatients.
Scandinavian journal of gastroenterology. Supplement (1989-01-01)
G Dobrilla, M Valentini, M C Bonoldi, L Piazzi
PMID2694331
RESUMO
Forty outpatients with endoscopically confirmed duodenal ulcers were entered in a double blind trial. They were randomly allocated to octatropine-methyl-bromide and sulglycotide salt (GVP) or placebo. The results show that the combination of the two drugs is less efficacious than the two constituent substances taken separately, is not more efficacious than placebo in ulcer healing, and is ineffective with regard to ulcer pain.
MATERIAIS