Merck
  • Novel hybrids of optically active ring-opened 3-n-butylphthalide derivative and isosorbide as potential anti-ischemic stroke agents.
Novel hybrids of optically active ring-opened 3-n-butylphthalide derivative and isosorbide as potential anti-ischemic stroke agents.
Journal of medicinal chemistry (2013-03-21)
Xiaoli Wang, Linna Wang, Tingting Li, Zhangjian Huang, Yisheng Lai, Hui Ji, Xiaolong Wan, Jinyi Xu, Jide Tian, Yihua Zhang
RESUMO
In search of novel anti-ischemic stroke agents with higher potency than a known drug 3-n-butylphthalide (NBP), a series of hybrids ((S)- and (R)-5a-f) from optically active ring-opened NBP derivative and isosorbide were synthesized for evaluating their anti-ischemic stroke activity. Compound (S)-5e displayed the strongest activity in inhibiting the adenosine diphosphate (ADP) and arachidonic acid (AA)-induced platelet aggregation in vitro, with 10.0- and 8.4-fold more effectiveness than (S)-NBP, respectively. Furthermore, (S)-5e was stable in artificial gastrointestinal fluids and could penetrate the blood-brain barrier (BBB) with an appreciate lipid/water partition coefficient relative to (S)-NBP. More importantly, oral treatment with (S)-5e protected from acute thrombosis and inhibited the ischemia/reperfusion-related brain injury in animals. Our findings suggest that (S)-5e may be promising for further evaluation for the intervention of ischemic stroke.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Dianhydro-D-glucitol, 98%
Preço e disponibilidade não estão disponíveis no momento.