Merck
  • The retail market for fresh cassava root tubers in the European Union (EU): the case of Copenhagen, Denmark--a chemical food safety issue?
The retail market for fresh cassava root tubers in the European Union (EU): the case of Copenhagen, Denmark--a chemical food safety issue?
Journal of the science of food and agriculture (2010-04-01)
Lotte Kolind-Hansen, Leon Brimer
RESUMO
A number of retail shops in Copenhagen sell fresh cassava roots. Cassava roots contain the toxic cyanogenic glucoside linamarin. A survey was made of the shop characteristics, origin of the roots, buyers, shop owner's knowledge of toxicity levels, and actual toxicity levels. Shops selling fresh cassava were shown mostly to be owned by persons originating in the Middle East or Afghanistan, buyers were found to predominantly be of African origin, and sellers' knowledge concerning the potential toxicity was found to be very restricted. Seventy-six per cent of the roots purchased had a total cyanogenic potentials (CNp) above the 50 mg HCN equivalents kg(-1) dry weight (d.w.) proposed as acceptable by an EU working group. Two of 25 roots purchased had CNp higher than 340 mg HCN eq. kg(-1) d.w. The EU has previously made risk assessments concerning cassava and cyanogenic compounds. In the light of the conclusions drawn, the EU needs to make decisions about how to deal with the regulation and control of fresh cassava roots imported to the European food market. Also cassava root products and cassava leaves should be considered.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
α-Hydroxyisobutyronitrile β-D-glucopyranoside, ≥97% (HPLC)