Merck
  • Transport, metabolism, and hepatotoxicity of flutamide, drug-drug interaction with acetaminophen involving phase I and phase II metabolites.
Transport, metabolism, and hepatotoxicity of flutamide, drug-drug interaction with acetaminophen involving phase I and phase II metabolites.
Chemical research in toxicology (2007-09-29)
Seva E Kostrubsky, Stephen C Strom, Ewa Ellis, Sidney D Nelson, Abdul E Mutlib
RESUMO
Treatment with flutamide has been associated with clinical hepatotoxicty. The toxicity, metabolism,and transport of flutamide were investigated using cultured human hepatocytes. Flutamide and its major metabolite, 2-hydroxyflutamide, caused an inhibition of taurocholate efflux in human hepatocytes with an IC50=75 microM and 110 microM, respectively. Treatment of hepatocytes with flutamide or 2-hydroxyflutamide for 24 h resulted in time- and concentration-dependent toxicity as assessed by inhibition of protein synthesis. Toxicity was greater after 1 h than after 24 h of treatment. Recovery in inhibition of protein synthesis by 24 h was attributed to the decreased presence of flutamide due to its metabolism. Flutamide was metabolized by hepatocytes to several metabolites, and formation of reactive intermediates of flutamide, as evidenced by the presence of glutathione-related adducts, was observed. Inhibition of flutamide metabolism by 1-aminobenzotriazole (ABT) resulted in enhancement of flutamide toxicity, which was associated with sustained levels of nonmetabolized drug. ABT also prevented the formation of reactive intermediates of flutamide. There was an additive toxicity when cells were treated with a combination of flutamide and 2-hydroxyflutamide. Simultaneous treatment with flutamide and acetaminophen (APAP) resulted in additive to synergistic toxic effects. Flutamide and APAP were found to have significant effects on each other's metabolism. Flutamide inhibited glucuronidation and sulfation of APAP, resulting in greater amounts of APAP available for bioactivation. APAP inhibited the hydroxylation of flutamide, and subsequent sulfation and acetylation of 4-nitro-3-(trifluoromethyl) aniline, a metabolite of flutamide. In summary, we suggest that inhibition of bile acid efflux by flutamide and its 2-hydroxy metabolite may play a role in flutamide-induced liver injury. Both flutamide and 2-hydroxyflutamide are responsible for cytotoxicity if not metabolized. The data also suggest a possible drug-drug interaction between flutamide and APAP, resulting in inhibition of flutamide metabolism and increased APAP bioactivation and toxicity.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Hydroxyflutamide, ≥98% (HPLC)