Merck
Nonaqueous capillary electrophoresis-mass spectrometry.
Journal of chromatography. A (2007-02-24)
Gerhard K E Scriba
RESUMO
Nonaqueous background electrolytes broaden the application of capillary electrophoresis displaying altered separation selectivity and interactions between analytes and buffer additives compared to aqueous background electrolytes. In addition, nonaqueous capillary electrophoresis (NACE) appears to be ideally suited for online coupling with mass spectrometry due to the high volatility and low surface tension of many organic solvents. Despite these advantages and an increasing use of nonaqueous background electrolytes in CE, coupling of NACE to mass spectrometry has not yet been applied very often to date. The present review summarizes the applications of online NACE-MS with regard to the analysis of drugs, stereoisomers, peptides, alkaloids, polymers and others. A brief discussion of solvent effects in NACE and pH of nonaqueous background electrolyte systems is also presented.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Nε-Trifluoroacetyl-L-lysine, ≥96.0% (TLC)