Merck
Biotin-decorated all-HPMA polymeric micelles for paclitaxel delivery.
Journal of controlled release : official journal of the Controlled Release Society (2020-09-15)
Yan Wang, Mies J van Steenbergen, Nataliia Beztsinna, Yang Shi, Twan Lammers, Cornelus F van Nostrum, Wim E Hennink
RESUMO
To avoid poly(ethylene glycol)-related issues of nanomedicines such as accelerated blood clearance, fully N-2-hydroxypropyl methacrylamide (HPMAm)-based polymeric micelles decorated with biotin for drug delivery were designed. To this end, a biotin-functionalized chain transfer agent (CTA), 4-cyano-4-[(dodecylsulfanylthiocarbonyl)-sulfanyl]pentanoic acid (biotin-CDTPA), was synthesized for reversible addition-fragmentation chain-transfer (RAFT) polymerization. Amphiphilic poly(N-2-hydroxypropyl methacrylamide)-block-poly(N-2-benzoyloxypropyl methacrylamide) (p(HPMAm)-b-p(HPMAm-Bz)) with molecular weights ranging from 8 to 24 kDa were synthesized using CDTPA or biotin-CDTPA as CTA and 2,2'-azobis(2-methylpropionitrile) as initiator. The copolymers self-assembled in aqueous media into micelles with sizes of 40-90 nm which positively correlated to the chain length of the hydrophobic block in the polymers, whereas the critical micelle concentrations decreased with increasing hydrophobic block length. The polymer with a molecular weight of 22.1 kDa was used to prepare paclitaxel-loaded micelles which had sizes between 61 and 70 nm, and a maximum loading capacity of around 10 wt%. A549 lung cancer cells overexpressing the biotin receptor, internalized the biotin-decorated micelles more efficiently than non-targeted micelles, while very low internalization of both types of micelles by HEK293 human embryonic kidney cells lacking the biotin receptor was observed. As a consequence, the paclitaxel-loaded micelles with biotin decoration exhibited stronger cytotoxicity in A549 cells than non-targeted micelles. Overall, a synthetic pathway to obtain actively targeted poly(ethylene glycol)-free micelles fully based on a poly(HPMAm) backbone was established. These polymeric micelles are promising systems for the delivery of hydrophobic anticancer drugs.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
N-Boc-1,6-hexanediamine, ≥98.0% (NT)