Merck
In vitro effects of conjugated linoleic acid (CLA) on inflammatory functions of bovine monocytes.
Journal of dairy science (2020-07-21)
G Ávila, C Catozzi, D Pravettoni, G Sala, P Martino, G Meroni, C Lecchi, F Ceciliani
RESUMO
The conjugated linoleic acid (CLA) isomers, a group of naturally occurring isomers of the essential fatty acid (FA) linoleic acid, have received special attention in animal and human nutrition. Although they have long been used as dietary integrators in dairy cows, the effects of CLA isomers on bovine immune cells remain mostly undisclosed. The present study aimed to cover this gap and investigate the in vitro effects of CLA on inflammatory functions, including chemotaxis, phagocytosis, killing capability, and extracellular respiratory burst of purified bovine monocytes (CD14+). The apoptosis rate of monocytes was addressed as well. Once assessed, the effects of different concentrations (10, 50, 100, and 500 μM) of the 2 main CLA isomers, namely cis-9,trans-11 and trans-10,cis-12, the experiments were carried out using a concentration of 50 μM of the CLA isomers, both individually and in a mixture (50:50). The immunomodulatory activities of linoleic acid, an essential FA, and stearic acid, a saturated FA, were also investigated. Only the 50:50 CLA mixture was able to reduce monocyte apoptosis and to increase the extracellular respiratory burst during experimental proinflammatory conditions, as assessed by measuring production of reactive oxygen species. Linoleic acid and CLA had no effects on chemotaxis, phagocytosis, or killing capability. Remarkably, treatment of monocytes with stearic acid significantly reduced their chemotactic capability. The present results demonstrated that CLA isomers do have immunomodulatory effects on some functions of bovine monocytes, and that the mixture of the 2 CLA isomers is more effective than the CLA isomers individually.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Triton X-100, laboratory grade
Large Column Holder, For columns o.d. 50 mm, ÄKTA®xpress, Cytiva 28400737