Merck
  • Thrombospondin-2 is upregulated in patients with aortic dissection and enhances angiotensin II-induced smooth muscle cell apoptosis.
Thrombospondin-2 is upregulated in patients with aortic dissection and enhances angiotensin II-induced smooth muscle cell apoptosis.
Experimental and therapeutic medicine (2020-10-24)
Liping Qi, Kui Wu, Shutian Shi, Qingwei Ji, Huangtai Miao, Que Bin
RESUMO
Thrombospondin-2 (TSP-2) is an important extracellular matrix protein that is involved in a variety of cardiovascular diseases, including viral myocarditis and abdominal aortic aneurysm. The present study aimed to investigate TSP-2 expression in patients with aortic dissection (AD). Aortas were obtained from patients with AD and healthy donors, and TSP-2 expression level in all samples was measured by western blotting and immunofluorescence assays. Blood samples were also collected from patients with AD and non-AD (NAD) subjects. Circulating TSP-2, tumor necrosis factor (TNF)-α and interleukin (IL)-6 levels in each sample were detected using ELISAs. In addition, the effect of TSP-2 on angiotensin II (Ang II)-induced smooth muscle cell (SMC) apoptosis was assessed in vitro. Compared with healthy donors, aortic TSP-2 expression level was significantly increased in patients with AD. Furthermore, TSP-2 was secreted primarily by SMCs, but also by endothelial cells. TSP-2 plasma expression level was also elevated in patients with AD compared with non-AD subjects. Furthermore, TSP-2 serum expression level was positively correlated with TNF-α and IL-6 expression levels in patients with AD. In addition, recombinant mouse TSP-2 treatment increased Bax mRNA expression and decreased Bcl2 mRNA expression in Ang II-treated SMCs; however, the effects were reversed following treatment with the NF-κB p65 signaling pathway inhibitor JSH-23 or with the anti-TNF-α and anti-IL-6 neutralizing antibodies. The present study demonstrated that TSP-2 expression was increased in the aortic tissues and plasma of patients with AD. These findings suggested that TSP-2 may participate in the progression of AD by activating the NF-κB p65 signaling pathway and amplifying the inflammatory response.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Indometacina, 98.5-100.5% (in accordance with EP)
Sigma-Aldrich
Angiotensin II, Human, Plays an important role in the regulation of blood pressure.