Merck
Critical role of the BAF chromatin remodeling complex during murine neural crest development.
PLoS genetics (2021-03-23)
Kathleen Wung Bi-Lin, Pratap Veerabrahma Seshachalam, Tran Tuoc, Anastassia Stoykova, Sujoy Ghosh, Manvendra K Singh
RESUMO
The BAF complex plays an important role in the development of a wide range of tissues by modulating gene expression programs at the chromatin level. However, its role in neural crest development has remained unclear. To determine the role of the BAF complex, we deleted BAF155/BAF170, the core subunits required for the assembly, stability, and functions of the BAF complex in neural crest cells (NCCs). Neural crest-specific deletion of BAF155/BAF170 leads to embryonic lethality due to a wide range of developmental defects including craniofacial, pharyngeal arch artery, and OFT defects. RNAseq and transcription factor enrichment analysis revealed that the BAF complex modulates the expression of multiple signaling pathway genes including Hippo and Notch, essential for the migration, proliferation, and differentiation of the NCCs. Furthermore, we demonstrated that the BAF complex is essential for the Brg1-Yap-Tead-dependent transcription of target genes in NCCs. Together, our results demonstrate an important role of the BAF complex in modulating the gene regulatory network essential for neural crest development.
MATERIAIS
Número do produto
Marca
Descrição do produto
Roche
In Situ Kit de detecção de morte celular, fluoresceína, sufficient for ≤50 tests, suitable for detection
Sigma-Aldrich
O9-1 Mouse Cranial Neural Crest Cell Line, stably expresses stem cell markers and neural crest markers