Merck
Biomolecular detection, tracking, and manipulation using a magnetic nanoparticle-quantum dot platform.
Journal of materials chemistry. B (2019-12-24)
Kalpesh D Mahajan, Gang Ruan, Greg Vieira, Thomas Porter, Jeffrey J Chalmers, R Sooryakumar, Jessica O Winter
RESUMO
Fluorescent and magnetic materials play a significant role in biosensor technology, enabling sensitive quantification and separations with applications in diagnostics, purification, quality control, and therapeutics. Here, we present a magneto-fluorescent biosensor/separations platform consisting of quantum dots (QDs) and superparamagnetic iron oxide nanoparticles (SPIONs) that are separately encapsulated in amphiphilic block co-polymer micelles conjugated to DNA or protein (i.e., single-stranded (ss) DNA derived from the mRNA of the tumor suppressor protein p53 or avidin protein). Analytes were detected via an aggregation sandwich assay upon binding of at least 1 QD and 1 SPION-containing micelle to result in a fluorescent/magnetic composite. Multiplexed isolation of protein and DNA biomolecules was demonstrated by using QDs of varying emission wavelength; QD fluorescence intensity could be correlated with analyte concentration. Sequential or parallel biomolecule separation was achieved by adding appropriately functionalized SPION-containing micelles and applying user-controlled magnetic fields via patterned magnetic disks and wires. QD fluorescence was used to continuously visualize analyte separation during this process. This QD/SPION platform is simple to use, demonstrates ∼10-16 M sensitivity in analyte detection (comparable to competing QD biosensors based on energy transfer) with specificity against 1 and 2 basepair mismatches in DNA detection, molecular separations capability in solutions of ∼10-10 M, and permits simultaneous or parallel, multiplexed separation of protein and DNA. Thus, this versatile platform enables self-assembly-based rapid, sensitive, and specific detection and separation of biomolecules, simultaneously and with real-time visualization. This technology demonstrates potential for nanoscale assembly, biosensing, and bioseparations.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
MES hemisodium salt, ≥98% (titration)
Preço e disponibilidade não estão disponíveis no momento.