Merck
  • Rhenium oxide nanoparticles - Sonochemical synthesis and integration on anode powders for solid oxide fuel cells.
Rhenium oxide nanoparticles - Sonochemical synthesis and integration on anode powders for solid oxide fuel cells.
Ultrasonics sonochemistry (2020-07-16)
Petros M Sakkas, Maria Argirusi, Georgia Sourkouni, Christos Argirusis
RESUMO
Rhenium oxide nanoparticles have been prepared using ultrasonication at 20 kHz. Samples characterization was committed via SEM-EDX, TEM, XRD, and Raman spectroscopy. Various experimental parameters were examined, including precursor/substrate amounts, ultrasonication intensity, and type of solvent used. Insights to the agglomeration of the prepared nanoparticles depending on the preparation parameters are given. As ultrasonic source we used either an ultrasonic probe by Sonics & Materials Inc. (20 kHz, 750 W net output) or a Bandelin SONOPULS HD 3200 ultrasound generator (20 kHz, 200 W net output) at intensities between 30 and 100 W/cm2. The rhenium oxide nanoparticles haven been decorated on state-of-the-art anode materials (NiO/GDC) for solid oxide fuel cells (SOFCs) in order to prepare catalytically more active anode powders. These experiments revealed that ultrasonication intensity and solvents used are able to affect final nanoparticles size distribution and morphology. At the same time, ratio of precursor and substrate compounds amounts as well as ultrasonication intensity and duration were all found to affect the decoration loading extend of nanoformations on substrate powders. The results showing the influence of the above-mentioned parameters allowed for the quantification of the effects on the loading and the preferable sites of the decoration.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Dirhenium decacarbonyl, 98%
Preço e disponibilidade não estão disponíveis no momento.