Merck
The interaction of covalently bound heme with the cytochrome c maturation protein CcmE.
The Journal of biological chemistry (2004-10-07)
Takeshi Uchida, Julie M Stevens, Oliver Daltrop, Edgar M Harvat, Lin Hong, Stuart J Ferguson, Teizo Kitagawa
RESUMO
The heme chaperone CcmE is a novel protein that binds heme covalently via a histidine residue as part of its essential function in the process of cytochrome c biogenesis in many bacteria as well as plant mitochondria. In the continued absence of a structure of the holoform of CcmE, identification of the heme ligands is an important step in understanding the molecular function of this protein and the role of covalent heme binding to CcmE during the maturation of c-type cytochromes. In this work, we present spectroscopic data that provide insight into the ligation of the heme iron in the soluble domain of CcmE from Escherichia coli. Resonance Raman spectra demonstrated that one of the heme axial ligands is a histidine residue and that the other is likely to be Tyr134. In addition, the properties of the heme resonances of the holo-protein as compared with those of a form of CcmE with non-covalently bound heme provide evidence for the modification of one of the heme vinyl side chains by the protein, most likely the 2-vinyl group.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Thrombin CleanCleave Kit