Merck
Encapsulation and Release of Functional Nanodroplets Entrapped in Nanofibers.
Small (Weinheim an der Bergstrasse, Germany) (2018-04-18)
Kusuma Thongchaivetcharat, Ratchapol Jenjob, Daniel Crespy
RESUMO
A method is presented for preserving the structural integrity of nanodroplets produced by emulsification. Droplets of different hydrophobic liquids such as essential oils or monomers are produced by the miniemulsion process. The miniemulsions are then electrospun to yield dextran nanofibers entrapping the hydrophobic nanodroplets. The nanodroplets are then successfully redispersed by dissolving the nanofibers in water. Furthermore, it is shown that nanofibers can be used to store a monomer and a catalyst as healing agents for ring-opening metathesis polymerization. After dissolution, the healing agents are released and a self-healing reaction takes place. Embedding, storage, and release of emulsion nanodroplets is a promising method that avoids potential destabilization of droplets by coalescence or Ostwald ripening.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Cyclohexanecarbonitrile, 98%