Merck
  • Neurokinin 1 receptor antagonists in the prevention of chemotherapy-induced nausea and vomiting: focus on fosaprepitant.
Neurokinin 1 receptor antagonists in the prevention of chemotherapy-induced nausea and vomiting: focus on fosaprepitant.
Future oncology (London, England) (2017-11-14)
Bernardo L Rapoport, Karin Jordan, Cindy Weinstein
RESUMO
Chemotherapy-induced nausea and vomiting (CINV) remains a challenge in cancer care. Improved understanding of CINV pathophysiology has triggered the development of new antiemetic therapeutic options, such as selective neurokinin-1 (NK1) receptor antagonists (RAs), which effectively prevent CINV when added to a standard antiemetic regimen (serotonin-3 RA and dexamethasone). Aprepitant and its water-soluble prodrug, fosaprepitant dimeglumine, are the most widely used NK1 RAs, with extensive clinical use worldwide. Recently, a Phase III trial prospectively evaluated fosaprepitant-based antiemetic therapy for CINV prevention in a large, well-defined nonanthracycline- and cyclophosphamide-based moderately emetogenic chemotherapy population. Fosaprepitant demonstrated significantly improved efficacy outcomes compared with a control regimen and was generally well tolerated, indicating that NK1 RAs are a valuable therapeutic option in this setting.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Fosaprepitant dimeglumine, ≥98% (HPLC)