Voltar
SGE Microvolume Syringe
Products
Categoria de produto
syringes (7)
Fabricante
Comprimento (mm)
DE (mm)
Volume (µl)
Compatibilidade
Forma da agulha
Tamanho do pacote
Medidor
Filtrar produtos
Showing 1-7 of 7
Comparar
Número do produto
Descrição
Preços
24411
SGE Microvolume Syringe,
5BR-7, Plunger -in-needle (8mm OD barrel), volume 5 μL, needle size 23 ga, needle L × O.D. 70 mm × 0.63 mm
Expandir
Ver preços
509221
SGE Microvolume Syringe,
1BR-5, Plunger-in-needle (8mm OD barrel), volume 1 μL, needle size 23 ga, needle L × O.D. 50 mm × 0.63 mm
Expandir
Ver preços
24408
SGE Microvolume Syringe,
1BR-7, Plunger -in-needle (8mm OD barrel), volume 1 μL, needle size 23 ga, needle L × O.D. 70 mm × 0.63 mm
Expandir
Ver preços
24401
SGE Microvolume Syringe,
0.5BR-7, Plunger -in-needle (8mm OD barrel), volume 0.5 μL, needle size 23 ga, needle L × O.D. 70 mm × 0.63 mm
Expandir
Ver preços
24400
SGE Microvolume Syringe,
0.5BNR-5, Plunger-in-needle (6.5mm OD barrel), volume 0.5 μL, needle size 23 ga, needle L × O.D. 50 mm × 0.63 mm
Expandir
Ver preços
509272
SGE Microvolume Syringe,
1BNR-OC-7/0.47, Plunger -in-needle (8mm OD barrel), volume 1 μL, needle size 26 ga, needle L × O.D. 70 mm × 0.47 mm
Expandir
Ver preços
509140
SGE Microvolume Syringe,
0.5BNR-5BV, Plunger -in-needle (6.5mm OD barrel), volume 0.5 μL, needle size 23 ga, needle L × O.D. 50 mm × 0.63 mm