Voltar
Replacement NMR needle with cone tip for eVol
®
electronic syringe
Products
Categoria de produto
needles (4)
Fabricante
Comprimento (cm)
DE (mm)
Filtrar produtos
Showing 1-4 of 4
Comparar
Número do produto
Descrição
Preços
Z687790
Replacement NMR needle with cone tip for eVol
®
electronic syringe,
for 500 μL, N25/500, L × O.D. 11.5 cm × 0.63 mm, gauge size 23
Expandir
Ver preços
Z687782
Replacement NMR needle with cone tip for eVol
®
electronic syringe,
for 50 μL, L × O.D. 18.0 cm × 0.63 mm, gauge size 23
Expandir
Ver preços
Z687766
Replacement NMR needle with cone tip for eVol
®
electronic syringe,
for 5 μL, L × O.D. 11.5 cm × 0.5 mm, gauge size 25
Expandir
Ver preços
Z687774
Replacement NMR needle with cone tip for eVol
®
electronic syringe,
for 50 μL, L × O.D. 11.5 cm × 0.5 mm, gauge size 25