Voltar
Pur-A-Lyzer
™
Floating Racks for Dialysis
Products
Categoria de produto
racks (4)
Tamanho do pacote
Filtrar produtos
Showing 1-4 of 4
Comparar
Número do produto
Descrição
Preços
PURXR
Pur-A-Lyzer
™
Floating Racks for Dialysis,
holds up to 7 Maxi Pur-A-Lyzer dialysis tubes, pack of 1 ea
Expandir
Ver preços
PURDR
Pur-A-Lyzer
™
Floating Racks for Dialysis,
holds up to 4 Midi Pur-A-Lyzer dialysis tubes, pack of 1 ea
Expandir
Ver preços
PURNR
Pur-A-Lyzer
™
Floating Racks for Dialysis,
holds up to 9 Mini and 4 Maxi Pur-A-Lyzer dialysis tubes, pack of 1 ea
Expandir
Ver preços
PURGR
Pur-A-Lyzer
™
Floating Racks for Dialysis,
holds 1 Mega Pur-A-Lyzer dialysis tube, pack of 1 ea