Voltar
Low Temperature Viscosity Standard
Products
Fabricante
Formato
Aplicação
Tipo de material de referência
Filtrar produtos
Showing 1-12 of 12
Comparar
Número do produto
Descrição
Preços
JF1L
Low Temperature Viscosity Standard,
JF1-L, viscosity 3.104 mPa.s (-20 °C), viscosity 6.089 mPa.s (-40 °C)
Expandir
Ver preços
N27B
Low Temperature Viscosity Standard,
ASTM D 2162 traceable
Expandir
Ver preços
N115B
Low Temperature Viscosity Standard,
ASTM D 2162 traceable
Expandir
Ver preços
N2B
Low Temperature Viscosity Standard,
ASTM D 2162 traceable
Expandir
Ver preços
P60W0C
Low Temperature Viscosity Standard,
ASTM D 2162 traceable
Expandir
Ver preços
JF1H
Low Temperature Viscosity Standard,
JF1-H, viscosity 4.885 mPa.s (-20 °C), viscosity 11.15 mPa.s (-40 °C)
Expandir
Ver preços
N120B
Low Temperature Viscosity Standard,
ASTM D 2162 traceable
Expandir
Ver preços
N480B
Low Temperature Viscosity Standard,
ASTM D 2162 traceable
Expandir
Ver preços
N14B
Low Temperature Viscosity Standard,
ASTM D 2162 traceable
Expandir
Ver preços
N1400B
Low Temperature Viscosity Standard,
ASTM D 2162 traceable
Expandir
Ver preços
N105B
Low Temperature Viscosity Standard,
ASTM D 2162 traceable
Expandir
Ver preços
J10
Low Temperature Viscosity Standard,
ASTM D 2162 traceable