2637301
Inlet Liner, Split/Splitless Type, Straight Design (packed with 10% OV-101 on 80/100 Chromosorb W HP)
pkg of 1 ea
About This Item
descrição
Split (wool packed)
packed
embalagem
pkg of 1 ea
fabricante/nome comercial
Varian 03-918956-00 (OEM)
C × D.E. × D.I.
54 mm × 5.0 mm × 3.4 mm
compatibilidade
configured for
for use with Varian 1078 Injector
Descrição geral
Deactivated (silanized) glass inlet liner for capillary injection ports.
- Fits Varian (1078 and 1079 Injector)
Escolha uma das versões mais recentes:
Certificados de análise (COA)