Documentos
22948
Supelco
Zero Volume Reducer 1/16 to 1/8 External
About This Item
Código UNSPSC:
41115700
Materiais
stainless steel
embalagem
pkg of 1 ea
conector
1/16 in. Valco (nut and ferrule included)
1/8 in. male Valco for 1/8 in. injector or detector
Produtos recomendados
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Choose from one of the most recent versions:
Certificados de análise (COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Atendimento ao cliente