Documentos Principais
Z742751
OrganoPlate® Perfusion Rocker Mini
About This Item
Código UNSPSC:
41121800
entrada CA/CC
110 - 230 V AC, universal plug set (Contains Euro, UK, NAmerica and Japan plug formats)
Descrição geral
The Perfusion Rocker Mini provides continuous medium flow in the OrganoPlate with minimal settings required. In contrast to standard continuous rockers the Perfusion Rocker Mini features optimized time settings and programs for use with the OrganoPlate.
The Perfusion Rocker Mini accomodates 2 stacks of plates and suitable for use in tissue culture incubators.
Medium perfusion (flow) in the OrganoPlate is established by passive leveling of liquids. The perfusion can be maintained indefinitely by placing the OrganoPlate on a MIMETAS Perfusion Rocker. This customized rocker platform fits easily in an incubator and enables interval rocking (switching sides every 5-15 minutes), as well as adjustable rocking angles.
Minimal settings are required for pump- and tube-free perfusion in OrganoPlates. Perfusion Rocker Mini features optimized time- and angle settings and programs for use with OrganoPlates.
Do I really need a Perfusion Rocker?
We strongly advise to use a MIMETAS Perfusion Rocker Mini to optimally control the perfusion flow in OrganoPlates. The Perfusion Rocker allows for detailed programming of an intermitting rocking action. For evaluation purposes a standard continuous rocker platform (see-saw) can be used. First check with your supplier if your rocker platform is suitable for use in cell culture incubators.
For some applications, for example neuronal cell culture, we advise not to perfuse the tissues. For these applications, a MIMETAS Perfusion Rocker is not required.
Informações legais
OrganoPlate is a registered trademark of MIMETAS B.V.
Perfusion Rocker is a trademark of MIMETAS B.V.
Escolha uma das versões mais recentes:
Certificados de análise (COA)
Visite a Biblioteca de Documentos
Artigos
Human renal proximal tubule epithelial cells (RPTECs) are commonly used to predict human renal drug permeability and to investigate drug efflux. We have generated transporter knockout (KO) cell lines using CompoZr® Zinc Finger Nuclease (ZFN) technology in a proprietary renal proximal tubule epithelial cell line.