Documentos Principais
Z660116
Bandelin SONOREX Ultrasonic bath accessories
insert basket for Z659037
Selecione um tamanho
1 EA
R$ 1.384,00
R$ 1.384,00
Previsão de entrega em01 de junho de 2025
Selecione um tamanho
Alterar visualização
1 EA
R$ 1.384,00
About This Item
Código UNSPSC:
41101500
NACRES:
NB.74
R$ 1.384,00
Previsão de entrega em01 de junho de 2025
Materiais
stainless steel
embalagem
pack of 1 ea
fabricante/nome comercial
Bandelin K 10 B
disponibilidade
not available in USA and Canada
C × L × P
250 mm × 190 mm × 50 mm
Descrição geral
During ultrasonic operation, nothing may be placed on the tank floor, as this could cause abrasion between the sonication material and the ultrasonic oscillating tank. The use of a suitable basket to hold the sonication material is therefore recommended. This stainless steel baskets is designed so that the sonication material is always in the optimal area of ​​the cavitation field (1-2 cm from the floor) in the oscillating tank. With the help of a basket, the sonication material can also be conveniently placed in the device and removed after operation, e.g. for rinsing, without the user coming into contact with the sonication liquid in the oscillating tank.
Características e benefícios
  • perfect positioning in the sound field
  • optimal loading and unloading
Informações legais
SONOREX is a trademark of Bandelin Electronic GmbH & Co. KG
Escolha uma das versões mais recentes:
Certificados de análise (COA)