Documentos Principais
Z659495
Bandelin SONOREX Ultrasonic bath accessories
positioning lid fits Z659460
About This Item
Código UNSPSC:
41103706
NACRES:
NB.74
Materiais
stainless steel
Características
for 2 beakers
embalagem
pack of 1 ea
fabricante/nome comercial
Bandelin DE 08
disponibilidade
not available in USA and Canada
Descrição geral
When using chemically aggressive liquids (e.g. TICKOPUR J 80 U), removing chemically aggressive or abrasive contamination or sonicating sample liquids, insert cups are used (indirect sonication). Because nothing may be placed on the bottom of the ultrasonic bath use a suitable perforated lids/insert strips to hold the insert cups. Several insert cups can be sonicated with different liquids at the same time.
Características e benefícios
  • For holding insert cups
  • Sound-absorbing
  • Shortening the heating phase
Informações legais
SONOREX is a trademark of Bandelin Electronic GmbH & Co. KG
Escolha uma das versões mais recentes:
Certificados de análise (COA)