About This Item
Código UNSPSC:
41103000
fabricante/nome comercial
Jencons
capacidade
3.5 L
externo A × L × P
280 mm × 300 mm × 200 mm
Descrição geral
Specially designed to provide a rugged container for long-term thermal storage of ice and dry ice on the bench. Smooth, easy-to-clean, chemically resistant white polystyrene body and lid have a 25 mm thick double insulated wall filled with expanded polystyrene foam. Made with four convenient support holes to accept your single channel pipettors.
Informações legais
Igloo is a registered trademark of Igloo Products Corp.
Escolha uma das versões mais recentes:
Certificados de análise (COA)