GN445835
Greiner 3-D Amino HTA™ plate
non-sterile, 1 test per plate, 25 ea
About This Item
Código UNSPSC:
41121800
embalagem
pack of 25 ea
fabricante/nome comercial
Greiner 445835
disponibilidade
available only in China
adequação
suitable for (cell culture)
Descrição geral
The HTA™multifunctional High-Throughput microArraying in Slide Format. 3-D Amino surface meets the needs of DNA, protein and tissue immobilisation. The HTA™ 1 can be used for the processing of one sample. The HTA™ 12 are partitioned into 12 flat compartments, each with a printable surface of 6×6 mm and a low well rim of 0.5 mm. Twelve samples can be processed simultaneously. Combine the advantages of the universal microplate platform with microarrays for diagnostic applications due to identical well geometry compared with a standard 96-well microplate.
Informações legais
HTA is a trademark of Greiner Bio-One GmbH
Escolha uma das versões mais recentes:
Certificados de análise (COA)